KPG stirrers with lateral blade
Supplier: Lenz Laborglas GmbH & CO.KG
06307201 06307010 06307003 06307301 06307303 06307305 06307311 06307001 06307005 06307008 06307011 06307013 06307015 06307205 06307208 06307210 06307211 06307213 06307215 06307310 06307400
6307201.EA 110 EUR
6307201. 6307010. 6307003. 6307301. 6307303. 6307305. 6307311. 6307001. 6307005. 6307008. 6307011. 6307013. 6307015. 6307205. 6307208. 6307210. 6307211. 6307213. 6307215. 6307310. 6307400.
KPG stirrers with lateral blade
Stirrers Stirring Tools
DURAN®, borosilicate glass with PTFE paddle and paddle safety securing screw. 10 mm shaft Ø. The length of the ground and polished bearing surface for the shaft Ø is 160 mm.
Order Now
SPECIFICATIONS
Environmentally Preferable