Counting chambers, Bürker
Supplier: GLASWARENFABRIK KARL HECHT
40443702 40443703
631-0923EA 51.3 EUR
631-0923 HECH40443703
Counting chambers, Bürker
Slides Counting Chambers
These counting chambers are suitable for blood tests and urine analysis.
  • 169 squares (0,05×0,05 mm) of 0,0025 mm² each for erythrocytes
  • 144 squares (0,2×0,2 mm) of 0,04 mm² each for leucocytes
  • Total area of ruling: 9,3 mm²
Order Now
SPECIFICATIONS
Environmentally Preferable