Staining jars
Supplier: GLASWARENFABRIK KARL HECHT
42450010 42461010 42460010
631-9313EA 29.4 EUR
631-9313 631-9310 631-9311
Staining jars
Jars Staining Jars
Soda-lime glass.
  • Three versions
  • For 16 or 10 slides
  • Acc. to Hellendahl or Schiefferdecker
Suitable for standard microscope slides (76×26 mm).