2-Nonanone, Supelco®

Supplier: Merck

Synonyms: Methyl heptyl ketone, Heptyl methyl ketone

MQ100

63969-1ML
63969-1MLEA 49 EUR
63969-1ML
2-Nonanone, Supelco®
2-Nonanone
Formula: C₉H₁₈O
MW: 142.24 g/mol
Boiling Pt: 192
Melting Pt: –15 °C
Density: 0.82 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009553
CAS Number: 821-55-6
EINECS: 212-480-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c8f751d89560efad8f183597b6ba48631c6a5aa&vid=5b16d4ab929242da6ab0bc5fd0f0a762ca1f2878&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR