Sulphuric acid 1 mol/l (2 N) aqueous solution BiopharmaceuticalProduction

Supplier: VWR Chemicals
Warning

6501.2500EA 69.8 EUR
6501.2500
Sulphuric acid 1 mol/l (2 N) aqueous solution BiopharmaceuticalProduction
Sulphuric acid
Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 3264
ADR: 8 III

Order Now

Specification Test Results

Titer 0.996 --> 1.004 mol/l
Appearance Passes test
Identification Passes test
Residual metal catalyst Unlikely to be present
Residual solvents (ICH Q3C) Excl.by manufact. process
Endotoxin Max. 2.5 EU/ml
Filtered through 0.2 μm Confirmed

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR