(-)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ≥99.0%, LiChropur™ for chiral derivatisation, Supelco®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ100

65363-100MG 65363-500MG
65363-100MGEA 131 EUR
65363-100MG 65363-500MG
(-)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ≥99.0%, LiChropur™ for chiral derivatisation, Supelco®
(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride

(R)-(-)-a-Methoxy-a-(trifluoromethyl)phenylacetyl chloride (Mosher’s acid chloride) is generally used as a chiral derivatizing agent.


Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.

Formula: C₁₀H₈ClF₃O₂
MDL Number: MFCD00044400
CAS Number: 39637-99-5
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=115a38bb0428c3335c3ae26375c050f17e53eea1&vid=db1074f44ade5281091e6964cf487824d930a412&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR