(+)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ≥99.0%, LiChropur™ for chiral derivatisation, Supelco®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ100

65365-100MG-F 65365-500MG-F
65365-100MG-FEA 139 EUR
65365-100MG-F 65365-500MG-F
(+)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ≥99.0%, LiChropur™ for chiral derivatisation, Supelco®
(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride

(R)-(-)-a-Methoxy-a-(trifluoromethyl)phenylacetyl chloride (Mosher’s acid chloride) is generally used as a chiral derivatizing agent.


Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.

Formula: C₁₀H₈ClF₃O₂
MDL Number: MFCD00067105
CAS Number: 20445-33-4
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f52c12e355b4497707c2905723aa09c047497022&vid=69da0e9ae264d2748152dd3dbf0014a243600059&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR