Titrator, Titronic® 300
Supplier: SI Analytics
285225810 285225820 285225800 285225830
661-0218EA 2660 EUR
661-0218 661-0219 661-0217 661-0220
Titrator, Titronic® 300
Titrators
The Titronic® 300 is a motor-driven burette for manual titration and and a precise dosing instrument for dispensable liquids, solvents and titrating agents. It can be used as a stand-alone instrument but it can also function as a titration and dosing burette in computer-controlled (automatic) titrating systems. Ideal for labs where high precision and flexibility are required.
Manual titration with hand controller (mouse):
Titration rate can be adjusted in five different steps to optimise titration speed and accuracy
Automatic calculation of results in different units and documentation on aMore Product Information