pH electrode calibration and maintenance solutions kit
Supplier: WTW
109415
662-0322EA 171 EUR
662-0322
pH electrode calibration and maintenance solutions kit
Buffers pH Calibration Buffers
Adapted to different electrode types for calibration and maintenance.
The pepsin cleaning solution is used to remove protein containing precipitation from diaphragms, the 3 molar KCl as a storage or refill solution.
Delivery information: Kit contains set of 250 ml each of pepsin cleaning solution; 3 M KCl solution and standard buffer solutions of pH 4,006, 6,865, 9,180.