Double-platinum electrode
Supplier: SI Analytics
285103512
662-1374EA 413 EUR
662-1374
Double-platinum electrode
Electrodes Other Electrodes
Double-platinum electrode, with glass shaft and two platinum pin sensors (Ø 1 mm). For detection of Bromine according to ASTM by potentiometric titration.