pH/mV/°C/ion meters, handheld, Seven2Go™ S2 / S8 Pro
Supplier: Mettler - Toledo
30207875 30207877 30207874 30207879 30207878 30207953 30207952 30207951 30207950 30207949
662-1845EA 1260 EUR
662-1845 662-1846 662-1844 662-1848 662-1847 662-1838 662-1837 662-1836 662-1835 662-1834
pH/mV/°C/ion meters, handheld, Seven2Go™ S2 / S8 Pro
Electrochemistry Meters pH and Multi-Parameter Meters
These robust meters provide durability and one-handed operation, making them ideally suited for mobile applications in the laboratory, at-line, or outdoors. Thanks to their IP 67 protection and drop test resistance, the devices are able to withstand harsh and demanding environments.
  • New intuitive menu guarantees ease of use
  • T-Pad with smart button arrangement enables easy and fast navigation
  • Instrument automatically recognises InLab ISM sensors and stores sensor ID, serial number, and last calibration data
  • Automatic or manual temperature compensation
More Product Information