pH/mV/°C meters, bench, SevenExcellence™ S400
Supplier: Mettler - Toledo
30046242 30046240 30046241 30092982
662-1467EA 3570 EUR
662-1467 662-1464 662-1466 662-2000
pH/mV/°C meters, bench, SevenExcellence™ S400
Electrochemistry Meters pH and Multi-Parameter Meters
Not only do these meters measure pH and ORP with classical sensors, but they also support pH measurement with ISFET technology. The instruments can effectively cope with complex applications and stringent requirements in regulated markets as well as with routine measurement tasks in laboratories.
  • Large colour display, user friendly touch screen and menu guidance in 10 languages
  • Flexible method concept (17 pre-defined and 50 user-defined methods) for high reproducibility and security
  • Fits three modules in any composition and order, can be expanded with
More Product Information