pH/mV/°C meters, bench top, Orion™ Versa Star Pro™

Supplier: Thermo Orion
VSTAR12 VSTAR10
662-2495EA 3240 EUR
662-2495 665-0284
pH/mV/°C meters, bench top, Orion™ Versa Star Pro™
Electrochemistry Meters pH and Multi-Parameter Meters
These bench top meters with conductivity module meet most challenging applications for pH, mV, ORP and temperature. The meters offer interchangeable measurement modules that allow multiple users to customise four separate channels to meet their specific requirements.

  • Colour display with adjustable text sizes; numeric keypad; onscreen measurement stability indicator
  • Auto-Read to hold a stable reading; ten password-protected methods per channel
  • Audible alarms automatically notify users when a set offset value, high/low limit value, and calibration time
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR