pH electrode, single pore glass
Supplier: HAMILTON BONADUZ
238160
662-9214EA 279 EUR
662-9214
pH electrode, single pore glass
Electrodes pH Electrodes
The Single Pore concept stands for precise, reliable, and fast readings with a proven, patented system that ensure optimal contact between the electrolyte and the sample.
  • Highest accuracy and fast response time thanks to patented Single Pore electrolyte contact
  • Wide applicability for example with emulsions, ion-weak media or general laboratory applications
  • Thanks to Single Pore clogging of the diaphragm is practically impossible
  • Refillable electrolyte Skylyte-CL
pH range: 0 to 14
Delivery information: Supplied with individual certificate and serial number.