Anti-UB Rabbit Polyclonal Antibody

Supplier: Merck Millipore (Oncogene)
662099-50
662099-50EA 631 EUR
662099-50
Anti-UB Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: UB (ubiquitin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR