pH/mV/ISE meter, handheld, Orion Star™ A324

Supplier: Thermo Orion
STARA3245 STARA3240
665-0276EA 2350 EUR
665-0276 665-0275
pH/mV/ISE meter, handheld, Orion Star™ A324
Electrochemistry Meters pH and Multi-Parameter Meters
This rugged, waterproof portable meter is designed for a wide range of pH, ion concentration, mV, ORP and temperature testing and field applications. It can be used in the most demanding locations thanks to its IP 67-rated housing. The meter offers one measuring channel.

  • Up to a five point pH calibration using automatic buffer recognition of NIST or DIN buffer groups
  • Up to a five point ISE calibration with selectable units of ppm, M, mg/L, %, ppb or none
  • Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
More Product Information
Order Now

SPECIFICATIONS

Model A324
Environmentally Preferable
pH range –2,000...+20,000
pH resolution 0,1/0,01/0,001
pH accuracy ±0,002
pH calibration Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
mV range ±2000,0
mV resolution 0,1
mV accuracy ±0,2 mV or ±0,05% of reading, whichever is greater
ISE range 0,0001 - 19900
ISE resolution 0,0001 minimum, 1 to 3 significant digits (user selectable)
ISE accuracy ±0,2 or ±0,05% of reading
ISE calibration Up to 5 points
ISE units ppm, M, mg/L, %, ppb, none
Temperature range –5,0...+105,0 °C
Temperature resolution 0,1 °C
Temperature accuracy ±0,1 °C
Temperature compensation Automatic or manual
Battery type 4 AA (included)
Battery life 800 h
Power supply AC adapter or 4 AA batteries
Compliance IP 67
Data logging - log function Manual / Automatic with AUTO-READ™
Data logging capacity 2000
Memory Non-volatile memory preserves data log, calibration log and meter settings
Languages English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
Connections BNC (pH, ORP or ion selective electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe)
Outputs RS232, USB
No. of channels 1
Calibration pH with calibration editing option, ion concentration (ISE) with calibration editing option, relative
Calibration points 1 point relative mV (RmV) mode, 1 point ORP mode
W×D×H 240×100×65 mm
Weight 0,5 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR