Diethyl ether ≥99.5%, AR®, Macron Fine Chemicals™

Supplier: Avantor Performance Materials

Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane

Stabilisation: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

6783-25EA 56.48 EUR
6783-25
Diethyl ether ≥99.5%, AR®, Macron Fine Chemicals™
Diethyl ether
Formula: (CH₃CH₂)₂O
MW: 74.12 g/mol
Boiling Pt: 35
Melting Pt: -123
Flash Pt: –40 °C
MDL Number: MFCD00011646
CAS Number: 60-29-7
Merck Index: 12,03852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) min 99.5%
E/E max. 0.1%
Residue after Evaporation max. 0.03%
Stabiliser (BHT) ∼5 ppm
Titrable Acid (meq/g) max. 0.0005
Water (KF) max. 0.2%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR