Tetrachlorosilane ≥99% (metals basis)

Supplier: Thermo Fisher Scientific
Danger

69182.AB 69182.AE 69182.K2 69182.L7
69182.ABEA 30.1 EUR
69182.AB 69182.AE 69182.K2 69182.L7
Tetrachlorosilane ≥99% (metals basis)
Tetrachlorosilane

Purity: 99% CAS No.: 10026-04-7 Molecular Formula: Cl4Si Formula Weight: 169.90 Physical Form: Liquid Density: 1.483 Appearance: Colourless Odour: Acrid Melting Point: -70° Boiling Point: 57.6° Merck Reference: 14,8500 UN No.: 1818 MDL No.: MFCD00011229

Formula: Cl₄Si
Boiling Pt: 57,6 °C
Melting Pt: –70 °C
Density: 1,483
Storage Temperature: Ambient
MDL Number: MFCD00011229
CAS Number: 10026-04-7
EINECS: 233-054-0
UN: 1818
ADR: 8,II
Merck Index: 14,8500

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR