N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (sulphobetaine-14, SB-14), Millipore®

Supplier: Merck Millipore Calbiochem

693017-100 693017-25 693017-5 693017-500GM
693017-100EA 4320 EUR
693017-100 693017-25 693017-5 693017-500GM
N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (sulphobetaine-14, SB-14), Millipore®
N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (Sulfobetaine-14, SB-14)
Formula: C₁₉H₄₁NO₃S
Storage Temperature: Ambient
MDL Number: MFCD00036910
CAS Number: 14933-09-6
EINECS: 239-003-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR