Supelco® Organic Reference Standard, Artemisinin

Supplier: Merck

69532-10MG
69532-10MGEA 77.9 EUR
69532-10MG
Supelco® Organic Reference Standard, Artemisinin
Standards Organic Standards
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9f32d9d9dd83675a08ef9a95817a29642dbc1f1&vid=c9f32d9d9dd83675a08ef9a95817a29642dbc1f1&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Synonym(s) Arteannuin; Qinghaosu
Environmentally Preferable
M-Clarity quality level MQ100
CAS No. 63968-64-9
MDL No. MFCD00081057
Molecular weight 282,33
Melting point 156 - 157 °C (lit.)
Shipping conditions STD
Storage conditions RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR