Potassium hydroxide 85,0-100,5, pellets NF, FCC, Macron Fine Chemicals™

Supplier: Avantor Performance Materials
Danger

Synonyms: Potash caustic, Caustic potash

6976-04EA 60.88 EUR
6976-04 6976-06 6976-28
Potassium hydroxide 85,0-100,5, pellets NF, FCC, Macron Fine Chemicals™
Potassium hydroxide

Pellets. GMP manufactured.

Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Density: 2.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1813
ADR: 8 II
REACH: 01-2119487136-33
Merck Index: 12,07806

Order Now

Specification Test Results

Meets F.C.C. Requirements
Meets N.F. Requirements
GMP Manufactured Product
Food GMP Manufactured Product
Storage: Preserve in tight containers. = 0.002 %
CAUTION: For Manufacturing, processing or repackaging 85.0 - 100.5 %
Bulk Pharmaceutical Chemical = 85.0 %
Bulk Food Chemical = 3.5 %
NF - Assay (Total Alkali) ≥85.0 %
NF - Carbonate (as K₂CO₃) ≤3.5 %
NF - Content of Potassium ≥59.9 %
NF - Limit of Sodium ≤1.0 %
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Insoluble Substances Passes Test
FCC - Assay (KOH) 85.0 - 100.5 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Insoluble Substances Passes Test
FCC - Lead (Pb) ≤2 mg/kg
FCC - Mercury (Hg) ≤0.1 ppm
FCC - Potassium Carbonate (K₂CO₃) ≤3.5 %
Solubility Passes Test
Appearance (white hygroscopic pellets)
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR