Nitrogen/protein analysers, Dumatherm®
Supplier: GERHARDT
14-0400
701-0261EA 0 EUR
701-0261
Nitrogen/protein analysers, Dumatherm®
Analysers Protein Analysers
For the analysis of solid and liquid samples in Dumatherm®, the sample is burned at high temperatures, followed by an analysis of the obtained gases.
  • Detection limit: 0,01 mg N absolute, absolute C 0,02 mg
  • Standard deviation up to 0,3% absolute with 150 mg EDTA
  • Automatic sample processing
  • Safe analysis and increased reproducibility productivity
  • Convenient control software
Determination of proteins in milk and dairy products, cereals, eggs and their products, meat and derivatives, malt and brewery raw materials, animal feed, beer, wine, brewery grain,More Product Information