Formaldehyde 37% in aqueous solution stabilised, BAKER, Laboratory Reagent, J.T.Baker®

Supplier: Avantor
Danger


Stabilisation: Stabilised with methanol

7041.9025EA 0 EUR
7041.9025 7041.9010 7041.91000
Formaldehyde 37% in aqueous solution stabilised, BAKER, Laboratory Reagent, J.T.Baker®
Formaldehyde
Formula: CH₂O
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
EINECS: 200-001-8
UN: 2209
ADR: 8,III

Order Now

Specification Test Results

Assay 37.0 - 38.0 %
Acidity passes test
Appearance of solution passes test
Identification passes test
Methanol (CH3OH) 9.0 - 15.0 %
Sulfated Ash max. 0.1 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR