Chlorine analyser, ChloroSense®
Supplier: Palintest
CS100
705-0650EA 1720 EUR
705-0650
Chlorine analyser, ChloroSense®
Analysers Elemental Analysers
The ChloroSense is designed to determine free and total chlorine in less than one minute.
  • Alternative to the DPD method ((N,N-diethylparaphenylenediamine)
  • Reagent-free and operator independent method
  • Patented sensor for fast and accurate results
Designed for drinking water, it is used in other applications including washing of food produce, testing of potable water storage tanks, pipe refurbishment as well as in hospitals.
Delivery information: Supplied with electrodes, calibration chip, sample bottles and carrying case.