Handheld colorimeter, DR900
Supplier: Hach
9385200
705-0744EA 3380 EUR
705-0744
Handheld colorimeter, DR900
Colorimeters Multiparameter Colorimeters
The DR900 is a robust multiparameter configured for immediate use.
  • Dustproof, waterproof and shockproof construction
  • Easy-to-use menu-driven step-by-step analysis
  • 90 pre-programmed methods
  • High-quality reagents are available for your applications. They are formulated to ensure exceptional performance and deliver results
Specially designed for field applications.
For a list of popular test kits for the handheld colorimeter, DR900 instrument please see the COMPLEMENTARY PRODUCTS Tab and for a more comprehensive selection of test kits available fromMore Product Information