BOD measurement, OxiTop®-i with graphic display
Supplier: WTW
208260 208262 208264 208266 208270 208272 208274 208276 208278
705-1144EA 4650 EUR
705-1144 705-1145 705-1148 705-1149 705-1150 705-1151 705-1152 705-1146 705-1153
BOD measurement, OxiTop®-i with graphic display
BOD Measurement Systems
The OxiTop®-i is a modern, easy to use instrument for BOD measurement (Biochemical Oxygen Demand). The duration can be defined between 1 and 7 days. The sample volume can be selected on the menu. The conversion into the corresponding BOD value will be automatic, error-free and convenient. A small LED show the actual operating condition of the head. The heads have a modern design, are available in two different colours with adjustable numeric sample ID allowing distinction between inlet and outlet.
  • No need to convert data - direct input of sample volume (select from 22,7 / 43,5 / 97,0 /
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable