CLASSIQSwabs™ Sterile Dry Fiber Wrapped Swabs
Supplier: Copan
164KS01 155C 160C 167KS01 167KS02 168C 170KS01 184CS01 185CS01 159C 155CS01 167KS100 175KS01 162C 167CS01 8155C 170KS01.US
710-0459EA 331 EUR
710-0459 710-0457 710-0430 710-0186 710-0460 710-0461 710-0187 710-0463 710-0464 710-0928 COPA155CS01 COPA167KS100 710-0931 710-0929 COPA167CS01 COPA8155C COPA170KS01.US
CLASSIQSwabs™ Sterile Dry Fiber Wrapped Swabs
Swabs Swabs for Sampling
CLASSIQSwabs™ are traditional and sterile ready-for-use devices intended for the collection of clinical samples.  The association of Copan CLASSIQSwabs™ with the wide range of Copan Transport Media enables an efficient sample collection and preservation compatible with many downstream assays.
These soft fiber swabs are available in tubes or peel pouches, Regular or Minitip shapes, and various materials such as cotton, rayon, and polyester. All these materials are inert and non-toxic to microorganisms and patients. CLASSIQSwabs™ are traditional and sterile ready-for-use devices intended
More Product Information