Tissue processing/embedding cassettes, Histosette® II, M485
Supplier: Simport Scientific
M485-11 M485-2 M485-3 M485-5 M485-6 M485-4 M485-10 M485-12 M485-7 M485-8 M485-9
720-0224EA 142 EUR
720-0224 720-0226 720-0227 720-0229 720-0230 720-0228 720-0223 720-0225 720-0231 720-0232 720-0233
Tissue processing/embedding cassettes, Histosette® II, M485
Embedding Cassettes Tissue Processing Cassettes
Acetal polymer, similar to M492, but covers are already removed from cassettes and are packed separately in the case.
  • Convenient for cassette labelling instruments as the covers are already removed
  • Resistant to the chemical action of histological solvents
  • Flow-through slots maximise fluid exchange