Biopsy processing/embedding cassettes, Swingsette®, M518
Supplier: Simport Scientific
M518-10 M518-11 M518-12 M518-2 M518-3 M518-4 M518-5 M518-6 M518-8 M518-9 M518-7
720-1670EA 197 EUR
720-1670 720-1671 720-1672 720-1676 720-1677 720-1678 720-1679 720-1680 720-1682 720-1683 720-1681
Biopsy processing/embedding cassettes, Swingsette®, M518
Embedding Cassettes Biopsy Processing Cassettes
Ideal for cassette-labelling instruments since the covers are packed separately in the case. These biopsy cassettes, made of acetal polymer, are similar to the M517 series but are especially designed to hold biopsy specimens during the processing/embedding process as well as in storage cabinets.
  • Moulded from a special high density polymer, they keep specimens safely submerged in liquid
  • Totally resistant to the chemical action of histological solvents
  • The efficient flow-through slots maximise fluid exchange and ensure proper drainage
  • The special hinge make
More Product Information