Wound and blister syringes with catheter fitting, Omnifix® Solo
Supplier: B. Braun
4613503F
720-2526EA 2.9 EUR
720-2526
Wound and blister syringes with catheter fitting, Omnifix® Solo
Syringes Syringes, Medical
PP, three-piece, single use, natural latex-free plunger stoppers with double sealing ring and highly transparent barrel.
  • Permanent, easy-to-read graduations
  • Safe plunger backstop
  • Centric catheter fitting
  • PVC- and latex-free