Sodium tetrakis(4-fluorophenyl)borate dihydrate ≥97.0%, Selectophore™ ionophore, Supelco®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ100

72014-500MG
72014-500MGEA 339 EUR
72014-500MG
Sodium tetrakis(4-fluorophenyl)borate dihydrate ≥97.0%, Selectophore™ ionophore, Supelco®
Sodium tetrakis(4-fluorophenyl)borate dihydrate

Sodium tetrakis(4-fluorophenyl)borate dihydrate is sodium salt derivative of tetraphenylborate (TPB).


Anionic additive Sodium tetrakis(4-fluorophenyl)borate dihydrate has been used during the potentiometric measurement of ammonia and blood urea nitrogen (BUN) sensors.


Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.

Formula: C₂₄H₂₀BF₄NaO₂
MDL Number: MFCD00149598
CAS Number: 207683-22-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50eb131437a63942ed399805e0898dd2f36ef501&vid=0cce6757d79f32391703b320a6418427173a781c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR