Sealing tape, light blocking, non-fluorescent, Nunc™
Supplier: Thermo Fisher Scientific
236703
731-0750EA 102 EUR
731-0750
Sealing tape, light blocking, non-fluorescent, Nunc™
Seals Microplate Seals
Vinyl film with pressure sensitive acrylate adhesive. For application to plate top or bottom, to protect light sensitive samples.
  • Temperature stable from −40 to +80 °C
  • Non-pierceable by pipette
Colour: black
External L×W: 123×79 mm