4-Nitrobenzoyl chloride, Supelco®

Supplier: Merck
Danger

Synonyms: p-Nitrobenzoyl chloride

M-Clarity™ quality level = MQ100

73120-100G
73120-100GEA 119 EUR
73120-100G
4-Nitrobenzoyl chloride, Supelco®
4-Nitrobenzoyl chloride
Formula: C₇H₄ClNO₃
MW: 185.57 g/mol
Boiling Pt: 278
Melting Pt: 97…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00007345
CAS Number: 122-04-3
EINECS: 204-517-4
UN: 3261
ADR: 8 II
Merck Index: 12,06687
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77311526916ced985e7c058e6a2144dd16d37886&vid=07672f7f9efcf84e6b7cecdbceb536def188f375&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR