PCR plates, 96-well, full skirt, Axygen®
Supplier: Corning
PCR-96-FS-C PCR-96-FS-CPBC
732-0664EA 58.9 EUR
732-0664 AXYGPCR-96-FS-CPBC
PCR plates, 96-well, full skirt, Axygen®
Microplates PCR Plates
PP plates, silicone mats. Designed to fit 0,2 ml thermal cycler blocks.
  • Ultra-thin walls of uniform thickness accelerate heat transfer, leading to shorter cycle times and more consistent results
  • Certified RNase, DNase and endotoxin-safe for highest sample integrity
  • AxyMat™ sealing mats autoclavable and reusable
Volume with AxyMat™: 140 μl
Compatible with (in addition to standard thermal cycling): BioRad iCycler, MyiQ, iQ5, Eppendorf Realplex, MJ Research Opticon, Chromo4, Stratagene MX3000.