Filter tips, ART®
Supplier: Thermo Fisher Scientific
2065 2065E 2069 2069G 2069IW 2070 2079E 2079EIW 2079GIW 2139 2139F 2140 2149 2149E 2149P 2149PIW 2159P 2160P 2180B 2279 2279IW 2780 2079 2065E-05 2069-05 2139-05 2139F-05 2140-05 2149P-05 2149E-05 2149P-50 2279-05
732-2202EA 77.1 EUR
732-2202 732-2204 732-2207 732-2210 732-2282 732-2212 732-2355 732-2217 615-0030 732-2219 732-2285 732-2221 732-2223 732-2293 732-2225 732-2301 732-2231 732-2233 732-2234 732-2236 732-2312 732-2238 732-2215 732-2205 732-2208 732-2220 732-2286 732-2222 732-2224 732-2294 732-2298 732-0995
Filter tips, ART®
Pipette Tips
Manufactured using a special process, tips are furnished with a unique ART® filter to eliminate any risk of cross-contamination between samples, which is particularly important in molecular biology. They are suitable for applications involving the use of DNA or RNA, as well as radioactive and infectious samples.
  • Certified free of RNase, DNase, DNA, ATP and pyrogens
  • Sterile tips: sterilised by gamma radiation
  • 'Low Retention' tips: Extremely hydrophobic surface for lower retention, superior precision and reproducibility of measurements