Eppendorf twin.tec® Real-Time PCR Plates, 96-Well, qPCR Plates
Supplier: EPPENDORF
0030132.505 0030132.513 0030132.530 0030132.548 0030132.700 0030132.718 0030129.644 0030129.636
732-2366EA 192 EUR
732-2366 732-2367 732-2369 732-2370 732-2372 732-2373 EPPE0030129.644 EPPE0030129.636
Eppendorf twin.tec® Real-Time PCR Plates, 96-Well, qPCR Plates
Microplates PCR Plates
Eppendorf twin.tec® real-time PCR plates, designed for low-volume qPCR, utilise white wells to enhance fluorescent light reflection, improving sensitivity and reproducibility in assays for reliable results.
  • White wells for better reflection
  • High mechanical stability
  • Effective sealing by raised well rims
  • Skirted and semi-skirted frames
  • Optimal heat transfer due to reduced wall thickness
  • Autoclavable (121 °C, 20 min)
  • Available in lot-certified PCR clean and Eppendorf Forensic DNA Grade for tested purity
The limiting factor in low-volume real-time PCR (More Product Information