VWR®, Filter Tips for Rainin® LTS® Pipettes, Low Retention
Supplier: VWR Collection
732-2896EA 153 EUR
732-2896 732-3339 732-3343
VWR®, Filter Tips for Rainin® LTS® Pipettes, Low Retention
Pipette Tips
These low retention filter tips are made with a special resin that reduces the potential for sample retention inside the tip. They help users reduce sample loss when performing protein research. These tips fit Rainin® LTS® pipettes.
  • Sustainable design based on virgin resins
  • Fit and eject with significantly less force than most conventional pipette tips
  • Ultra-fine points and super-clear resin for excellent viewing of sample
  • Free of RNase, DNase, endotoxins, human DNA and trace metals
  • Lot certified to be free of protease and nuclease
  • Tested for ATP