VWR® Rigid qPCR Plates, 96-Well, with White Wells
Supplier: VWR Collection
732-4403EA 66.1 EUR
732-4403 732-4405
VWR® Rigid qPCR Plates, 96-Well, with White Wells
Microplates PCR Plates
Rigid qPCR Plates feature a rugged polycarbonate frame, providing a solid and flat surface that guarantees seamless interaction with automated liquid handling platforms. With a firm grip assured, robots can confidently place these plates with utmost precision, eliminating any concerns of misalignment.
  • Polycarbonate frame is rigid and flat, facilitating a firm grip by robotic arms and ensuring reliable placement
  • Polypropylene wells provide optimal thermal transfer and higher reaction efficiency
  • White wells enhance fluorescence signals during qPCR
  • Wells are
More Product Information