VWR®, Taq DNA polymerase

Supplier: VWR Chemicals

733-1300EA 62.7 EUR
733-1300 733-1301 733-1302 733-1303 733-1305 733-1307 733-1312 733-1313 733-1819 733-1820 733-1823 733-2009
VWR®, Taq DNA polymerase
Nucleic Acid Reagents End-point PCR Enzymes and Kits

VWR® Taq DNA Polymerase is an ultra-pure, thermostable, recombinant DNA polymerase, which provides robust PCR performance in a wide range of PCR applications, without time-consuming optimisation. The enzyme is isolated from Thermus aquaticus, and has a molecular weight of approximately 94 kDa. VWR® Taq DNA Polymerase has both a 5' to 3' DNA polymerase and a double strand 5' to 3' exonuclease activity. It leaves an A overhang, which makes the enzyme ideal for TA cloning.


  • Ideal choice for routine applications
  • High performance, thermostable DNA polymerase
  • Optimal for TA cloning


Taq DNA polymerase concentration: 5 Units/μl


10X Key Buffer: Tris-HCl pH 8,5; (NH₄)₂SO₂, 15 mM MgCl₂, 1% Tween® 20
10X Extra Buffer: Tris-HCl pH 8,3; KCl, 15 mM MgCl₂, 1% Triton™ X-100
10X Mg-Free Key Buffer: Tris-HCl pH 8,5; (NH₄)₂SO₂, 1% Tween® 20
10X Mg-Free Extra Buffer: Tris-HCl pH 8,3; KCl, 1% Triton™ X-100


EU = Units


Delivery information: VWR® Taq DNA Polymerase is usually supplied with either or both Key Buffer and Extra Buffer. Key Buffer (NH⁴⁺) gives a superior amplification signal (high yield) minimising the need for optimisation of the Mg²⁺ concentration, or the annealing temperature in most primer-template systems. Extra Buffer is a traditional potassium (K⁺) buffer. Extra Buffer promotes high specificity, but careful optimisation of primer annealing temperatures and Mg²⁺ concentrations may be required.

Order Now

SPECIFICATIONS

Description Taq DNA Polymerase (733-1300) Taq DNA Polymerase (733-1301) Taq DNA Polymerase (733-1302) Taq DNA Polymerase (733-1303) Taq DNA Polymerase (733-1305) Taq DNA Polymerase (733-1307) Taq DNA Polymerase (733-1312) Taq DNA Polymerase (733-1313) Taq DNA Polymerase (733-1819) Taq DNA Polymerase (733-1820) Taq DNA Polymerase (733-1823) Taq DNA Polymerase (733-2009)
Environmentally Preferable
Format Enzyme with/without buffer Enzyme with/without buffer Enzyme with/without buffer Enzyme with/without buffer Enzyme with/without buffer Enzyme with/without buffer Enzyme with/without buffer Enzyme with/without buffer Enzyme with/without buffer Enzyme with/without buffer Enzyme with/without buffer Enzyme with/without buffer
Features Detergent free Detergent free Mg-free Mg-free Detergent free Mg-free
Hot start No No No No No No No No No No No No
Concentration 5 U/µl 5 U/µl 5 U/µl 5 U/µl 5 U/µl 5 U/µl 5 U/µl 5 U/µl 5 U/µl 5 U/µl 5 U/µl 5 U/µl
Units 250 500 1000 10000 1000 1000 1000 2500 2500 5000 10000 10000
Size 1 x 0.05 ml + 3 x 1.5 ml 1 x 0.1 ml + 3 x 1.5 ml 2 x 0.1 ml + 6 x 1.5 ml 3 x 0.667 ml + 18 x 5 ml 2 x 0.1 ml + 4 x 1.5 ml 2 x 0.1 ml + 4 x 1.5 ml 2 x 0.1 ml + 4 x 1.5 ml 5 x 0.1 ml + 10 x 1.5 ml 5 x 0.1 ml + 15 x 1.5 ml 10 x 0.1 ml + 9 x 5 ml 3 x 0.667 ml + 18 x 5 ml 3 x 0.667 ml + 12 x 5 ml
Buffer composition 10x Key Buffer, 10x Extra Buffer and 25 mM MgCl2 10x Key Buffer, 10x Extra Buffer and 25 mM MgCl2 10x Key Buffer, 10x Extra Buffer and 25 mM MgCl2 10x Key Buffer, 10x Extra Buffer and 25 mM MgCl2 10x Extra Buffer and 25 mM MgCl2
Detergent free
10x Key Buffer and 25 mM MgCl2
Detergent free
10x Key Buffer Mg-free and 25 mM MgCl2 10x Key Buffer Mg-free and 25 mM MgCl2 10x Key Buffer, 10x Extra Buffer and 25 mM MgCl2 10x Key Buffer, 10x Extra Buffer and 25 mM MgCl2 10x Key Buffer, 10x Extra Buffer and 25 mM MgCl2
Detergent free
10x Key Buffer Mg-free and 25 mM MgCl2
High fidelity No No No No No No No No No No No No
Amplicon size ≤ 5kb ≤ 5kb ≤ 5kb ≤ 5kb ≤ 5kb ≤ 5kb ≤ 5kb ≤ 5kb ≤ 5kb ≤ 5kb ≤ 5kb ≤ 5kb
Processivity Standard Standard Standard Standard Standard Standard Standard Standard Standard Standard Standard Standard
Exonuclease activity 5’→3’ 5’→3’ 5’→3’ 5’→3’ 5’→3’ 5’→3’ 5’→3’ 5’→3’ 5’→3’ 5’→3’ 5’→3’ 5’→3’
Product overhang 3'A 3'A 3'A 3'A 3'A 3'A 3'A 3'A 3'A 3'A 3'A 3'A
Storage conditions – 20 °C (long term), + 4 °C (6 months) – 20 °C (long term), + 4 °C (6 months) – 20 °C (long term), + 4 °C (6 months) – 20 °C (long term), + 4 °C (6 months) – 20 °C (long term), + 4 °C (6 months) – 20 °C (long term), + 4 °C (6 months) – 20 °C (long term), + 4 °C (6 months) – 20 °C (long term), + 4 °C (6 months) – 20 °C (long term), + 4 °C (6 months) – 20 °C (long term), + 4 °C (6 months) – 20 °C (long term), + 4 °C (6 months) – 20 °C (long term), + 4 °C (6 months)
Application Standard amplification Standard amplification Standard amplification Standard amplification Standard amplification Standard amplification Standard amplification Standard amplification Standard amplification Standard amplification Standard amplification Standard amplification

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR