Cell strainers, Falcon®
Supplier: Corning
352340 352350 352360
734-0002EA 143 EUR
734-0002 734-0003 734-0004
Cell strainers, Falcon®
Cell Strainers
An easy, ready-to-use way to consistently obtain a more uniform single-cell suspension. Made of a strong nylon mesh and available with 40, 70 or 100 µm pores that are evenly spaced for optimal performance in a variety of applications. A faster and easier alternative to gauze filtration in procedures involving dissociation of cells from either clumps or primary tissue.
  • Moulded, colour-coded PP frame with tab enables easy handling
  • Fits perfectly into a 50 ml Falcon® conical tube or other similarly sized tube
  • Sterilised by gamma irradiation and conveniently
More Product Information