Cover glasses, Corning® BioCoat™
Supplier: Corning
354089 354085 354086 354087 354088
734-1009EA 276 EUR
734-1009 734-1005 734-1006 734-1007 734-1008
Cover glasses, Corning® BioCoat™
Cover Glasses
These cover glasses have a uniform coating of either poly-L-lysine, poly-D-lysine and laminin, human fibronectin or rat tail collagen.
  • Optically clear
  • Low background fluorescence
  • Non-neurotoxic
Designed for applications where optically clear, thin glass bottoms are needed such as immunofluorescence microscopy, fluorescence imaging of live cells, confocal microscopy, high resolution and inverted microscopy and phase contrast microscopy.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 14001 - Bedford