Microplates, 24-well, Whatman™ UNIFILTER®
Supplier: Whatman products (Cytiva)
7700-9901 7700-9904 7700-9917
77009901.EA 1260 EUR
77009901. 734-2550 734-2551
Microplates, 24-well, Whatman™ UNIFILTER®
Microplates Filter Plates
The 10 ml UNIFILTER is used with large sample or reagent volumes. Typical applications include biomolecule purification by solid phase extraction and organic synthesis in combinatorial chemistry library generation.
  • Polypropylene microplate is heat and chemical resistant
  • 24 well microplate with 10 ml wells appropriate for large sample volumes
  • No cross talk: patented integral filter drip director design prevents well-to-well cross contamination
  • Filtration media options include glass microfiber, melt-blown polypropylene, PTFE, VFE
Applications such asMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable