VWR®, Multilayer Cell Culture Flasks, 3 Layer, 5 Layer
Supplier: VWR Collection
734-3415EA 160 EUR
734-3415 734-3416 734-3417 734-3418
VWR®, Multilayer Cell Culture Flasks, 3 Layer, 5 Layer
Flasks Cell Culture Flasks
Multilayer cell culture flasks enable you to grow more cells faster and easier, thereby making your cell culture workflow more productive.
  • Even distribution of media across all layers for homogenous cell growth
  • Flexible design allows to pour or aspirate/recover cells using a pipet
  • Consistent surface treatment for predictable scale-up
  • Molded graduations stand out distinctly on these optically clear
  • DNase/RNase free and non-pyrogenic
  • Sterilized by irradiation SAL 10⁻⁶ (ISO 11137)
  • Shelf Life: 3 years after month of production
Vent caps incorporate a 0,22More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 14001/Certificate No.: 04923E00243R0M