Organic reference standard, Indigo carmine, analytical standard, Supelco®
Supplier: Merck
73436-25MG
73436-25MGEA 208 EUR
73436-25MG
Organic reference standard, Indigo carmine, analytical standard, Supelco®
Standards Organic Standards
Indigo carmine is a synthetic indigoid dye, which is generally used as a food colouring agent. It is basically used to maintain the natural color of food during processing or storage and also to improve the color, texture and appearance of food products.
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1537f6ea888afbbdfcca28d079b56d84e2e469a8&vid=1537f6ea888afbbdfcca28d079b56d84e2e469a8&prid=web&clid=SAPDAM