Non-Contact Dispensers, I.DOT
Supplier: DISPENDIX
D16110022091 D16110024910 D16110026000 D16110021796 D16110021802 D16110021808 D16110021814 D16110024973
735-0642EA 128530 EUR
735-0642 735-0643 735-0644 735-0646 735-0647 735-0648 735-0649 735-0650
Non-Contact Dispensers, I.DOT
Automated Liquid Handling Systems
An accessible research appliance enabling the dispensing of nanoliter and microliter scale volumes to 96-, 384-, and 1536-well plates.
  • Use a fraction of the pipette tips required by traditional automation
  • Dispense reagents with a 1 μl dead volume and run more samples for the same reagent cost
  • Dispense 4 nl to 30 μl into any SBS format destination labware with a high degree of precision
  • Dispense with insight into the actual volume dispensed every time with integrated volume verification
  • Software for dispensing a constant volume, dilution, normalization, and
More Product Information