96-Well microplates, non treated, BRANDplates®
Supplier: Brand
781600 781601 781602 781603 781604 781605 781607 781608 781610 781611 781660 781661 781662 781663 781665 781668 781670 781671 781614 781615
735-2000EA 136 EUR
735-2000 735-2001 735-2002 735-2003 735-2004 735-2005 735-2006 735-2007 735-2008 735-2009 735-2022 735-2023 735-2024 735-2025 735-2026 735-2027 735-2028 735-2029 612-3668 738-0190
96-Well microplates, non treated, BRANDplates®
Microplates HTS Microplates
PS, solid plates or with transparent base. Plates are available with different well bottom shapes (round U-bottom, conical V-bottom, flat F-bottom or C flat bottom with curved edges), in different colours, and either sterile or non sterile.
  • All BRANDplates® are alphanumerically labelled, stackable and compatible with most standard microplate readers and washers
  • White plates provide maximum reflection and minimum crosstalk during luminescence measurements; black plates provide minimum back light scatter during fluorescence analysis
  • Sterile plates are
More Product Information