Deepwell Plates 96-well, 500 µl
Supplier: EPPENDORF
0030502.132 0030501.110 0030501.136 0030502.108 0030502.116 0030502.140 0030505.107 0030501.144 0030506.103 0030501.101 0030113.543
737-2518EA 445 EUR
737-2518 737-2501 737-2503 737-2515 737-2516 737-2519 737-0200 737-2504 737-0203 737-2500 737-0252
Deepwell Plates 96-well, 500 µl
Microplates Deep Well Microplates
Eppendorf Deepwell plates 96-well, 500 µl offer efficient sample handling with features like OptiTrack® matrix for rapid identification and g-Safe® technology for stability during centrifugation. SafeCode variants provide secure three-level coding, including error correction for reliable identification, ensuring flexibility and accuracy in experimental workflows.
  • OptiTrack® matrix: up to 30% faster sample identification and fewer pipetting errors via high-contrast alphanumeric labeling
  • RecoverMax® well design: Optimised well geometry for maximum sample
More Product Information