Eppendorf® Microplates
Supplier: EPPENDORF
0030601.106 0030601.203 0030601.300 0030602.102 0030602.200 0030602.307 0030606.302 0030621.107 0030621.301 0030622.103 0030622.308 0030113.586 0030113.594
737-0166EA 249 EUR
737-0166 737-0167 737-0168 737-0173 737-0174 737-0175 737-0224 737-0225 737-0226 737-0229 737-0230 737-0254 737-0255
Eppendorf® Microplates
Microplates HTS Microplates
Eppendorf microplates provide exceptional stability during centrifugation with g-Safe® technology and efficient sample identification with the OptiTrack® Matrix. The design features, including RecoverMax® well design and raised well rims, ensure reliable sealing and facilitate maximum sample recovery.
  • g-Safe®: exceptional centrifugation stability up to 6000×g
  • OptiTrack® Matrix: up to 30% faster sample identification and fewer pipetting errors via high-contrast alphanumeric labelling
  • RecoverMax® well design: Optimised well geometry for minimal remaining
More Product Information