Supelco® TraceCERT® Organic Reference Standard, Benz[a]anthracene
Supplier: Merck
75451-50MG
75451-50MGEA 116 EUR
75451-50MG
Supelco® TraceCERT® Organic Reference Standard, Benz[a]anthracene
Standards Organic Standards
  • GHS signal: GHS08, GHS09
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Caution: P201 - P273 - P308 + P313
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=560ff793e397597f25569c3d37edd5a47c889175&vid=560ff793e397597f25569c3d37edd5a47c889175&prid=web&clid=SAPDAM