J.T.Baker® BAKERBOND®, Ion Exchange Chromatography Resin, PolyQUAT Multimode
Supplier: Avantor
7603-01EA 208.21 EUR
7603-01 7603-02 7603-03 7603-04 7603-05
J.T.Baker® BAKERBOND®, Ion Exchange Chromatography Resin, PolyQUAT Multimode
Chromatography Sorbents HPLC Sorbents
The J.T.Baker® BAKERBOND® PolyQUAT multimode ion exchange (IEX) chromatography resin functions as a strong anion exchange resin with improved selectivity for superior separation in process chromatography applications.
Powered by a proprietary surface chemistry combining strong anion exchange functionality with weak anionic exchange sites and hydrophilicity, BAKERBOND® PolyQUAT multimode cation exchange chromatography media utilizes its unique selectivity to enable higher purification performance across a wide range of biopharmaceuticals.
Designed and manufactured by Avantor to the high standards established for our J.T.Baker® brand chemicals, BAKERBOND® PolyQUAT ion exchange resin can be used with other J.T.Baker® BAKERBOND® resins to enhance the separation power of your chromatography process steps.
Ordering information: 1 ml and 5 ml lab columns are also available under complementary products.